Would you like to log in again?
Pine Life Cycle Riker Mount, Ward’s®
Supplier: Riker Mount
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
R148
470128-622EA 73.1 USD InStock
470128-622
Pine Life Cycle Riker Mount, Ward’s®
Specimens Preserved Specimens Display Specimens
Explore gymnosperm life cycles.
  • Real specimens
  • Easy to store
This exhibit explores the life cycle and stages of the Pine (Pinus sp.). Stages include a male pollen cone, a first-year female cone, a mature female cone, a winged seed, a seedling, and the needle from a mature tree. Each specimen is fully labeled in a sealed display.
Ordering information: As these are prepared from real specimens, the product you receive may appear different from the photograph.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty