Would you like to log in again?
Crustacean Comparison, Riker Mount
Supplier: WHITE OWL ENTERPRISES
R225
470128-630EA 86.95 USD InStock
470128-630
Crustacean Comparison, Riker Mount
Specimens Preserved Specimens Display Specimens
Identify or compare commonly found crustaceans.
  • Real specimens
  • Wet and dry preservation
  • Sealed box
Class Crustacea is well-represented by this set of ten specimens. Each specimen is labeled, and all come housed in a display.
Ordering information: As these are prepared from real specimens, the product you receive may appear different from the photograph.
Delivery information: Included are the Decapods (Crab, Mole Crab, Crayfish, Grass Shrimp, and Shrimp), Isopods (Wood Louse), Amphipods (Sand Flea and Skeleton Shrimp), and Cirripeds (Goose Barnacle and Acorn Barnacle).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty