Would you like to log in again?
Ward's® Drosophila Culturing Kit
Supplier: VWR
470129-020KT 175.79 USD InStock
470129-020
Ward's® Drosophila Culturing Kit
Specimens Live Specimens Animals - Living Material
Observe the complete life cycle of Drosophila melanogaster, a widely used model for genetics research.
  • Observe mating, egg laying, larvae development, pupation, and emergence of adults
  • Perform a simple dominance cross
  • Flies provided have easily visable characteristics
This kit includes:
Coupon for 2 of each of the following fly strains: Vestigial and Wild-type
Anesthetizer and ice pack to sedate flies
Drosphila vials, caps, and media
Brush to transfer anesthetized flies
Instructions
Needed but not included:
Magnifying glass or dissection microscope
Ordering information: This item contains a coupon for live or perishable materials. At checkout, please provide the date you wish the perishable components to arrive.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground. Perishable components will ship via overnight or second day delivery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty