Would you like to log in again?
TeacherGeek Advanced Ping-Pong ball launcher
Supplier: TeacherGeek
91100 1823-05
470132-380EA 74.49 USD InStock
470132-380 470132-378
TeacherGeek Advanced Ping-Pong ball launcher
Educational Materials Physics Educational Materials TeacherGeek System Components
Includes more parts than the basic projectile launcher
  • Easy to connect parts
  • Reusable pieces
  • Hands on way to investigate the design process
  • A true STEM experience
The Ping-Pong Ball Launcher combines the two favorites, building and making things fly. This activity comes with all the components to build an adjustable launcher base and a launcher mechanism. From there choose a design purpose. Do you want to hit a target at a given distance? Do you want to launch the ping pong ball the highest or farthest? Is consistency important? How do the constants or variables affect your results (angle of trajectory, gravity, stored energy, mass, range, environment, etc.)?
Now design, build, implement and innovate with your launcher to allow for experimentation with the elements of projectile motion. This advanced activity includes additional parts and components that allow your design and innovation to go way beyond the basic launcher. How about building a catapult, ram, sling shot, kicker or whatever else you can imagine to launch the ball and achieve your goals.
The Launcher Activity Teaches...
Newton's Laws
Kinematics
Two-Dimensional Motion
Forces
Conservation of Energy
Simple Machines
Data Collection
Data Analysis
Construction
Design and Innovation
Engineering Challenges
Use kinematic equations for motion and two dimensions to determine the range of a projectile.
Determine which launcher angle resulted in the greatest distance traveled.
Determine which launcher angle results in consistently hitting a given target.
What forces act on a projectile in flight?
How does the mass of the projectile affect the distance it will travel when the same force and launch angle are used?
Calculate the average distance traveled for each angle.
Calculate the standard deviation of the distance traveled for each angle.
Create a spreadsheet / graph / table of your results.
Give a presentation using your data as argumentation for your best design.
Ordering information: Require but not included are Reamer, Easy Cutter, Phillips Screwdriver and Slip-Joint Pliers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty