Would you like to log in again?
TeacherGeek Wind Lift
Supplier: TeacherGeek
1823-14 1823-15
470132-398EA 11.95 USD InStock
470132-398 470132-400
TeacherGeek Wind Lift
Educational Materials Physics Educational Materials TeacherGeek System Components
Lift Weight and Mass with the Power of Wind
  • Easy to connect parts
  • Reusable pieces
  • Hands on way to investigate the design process
  • A true STEM experience
  • Build stators and coils
The TeacherGeek Wind Lift Activity allows you to build a wind powered lift. You will start by creating the stand from a hole plate, dowels, perpendicular blocks and hardware. Then the lift mechanism, hub and blades are added. Easy to follow pictorial steps are found in a free downloadable document. Encouraging tips are offered for experimentation with blade designs that lead to a variety of outcomes. Check the Wind Lift Activity Guide online for further details (available as a free download).
Engineering Challenges
Design at least four different style blades with varying shape, contour, length and number (2,3 or 6). Test each one to see which can lift the most weight.
Prepare a spreadsheet to record distance, time and weight. Calculate the horsepower of each design to determine which can produce the most work.
Ordering information: Required but not included are Reamer, Easy Cutter, Phillips Screwdriver and Slip-Joint Pliers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty