Would you like to log in again?
TeacherGeek Pulley Set - 40-PIECE mixed colors
Supplier: TeacherGeek
1823-29
470132-426EA 21.9 USD InStock
470132-426
TeacherGeek Pulley Set - 40-PIECE mixed colors
Educational Materials Physics Educational Materials TeacherGeek System Components
Pulley set to expand your TeacherGeek ideas.
The TeacherGeek 40 Piece Multi-Color Pulley Set contains ten each of the following sizes: 70mm, 55.5mm, 25.5mm, 9mm . These are used to create pulley driven transmissions such as for vehicles, wind turbines, lifts and pumps. Splined holes hold the dowels for a secure fit. Also the off center holes may be used to incorporate cams into the design. The unique pulley design allows for non-slip grip and easy exchange of rubber bands.
Multi-Color Set includes:
10 - 70mm pulleys
10 - 55.5mm pulleys
10 - 25.5mm pulleys
10 - 9mm pulleys
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty