Would you like to log in again?
TeacherGeek Perpendicular Block Pack of 100
Supplier: TeacherGeek
1823-34
470132-436EA 13.95 USD InStock
470132-436
TeacherGeek Perpendicular Block Pack of 100
Educational Materials Physics Educational Materials TeacherGeek System Components
Bulk perpendicular connectors to expand your TeacherGeek collection
The versatile TeacherGeek Perpendicular Block is an essential component for building most any activity. There are three splined holes that can be used to connect dowels at 90 degrees. The holes can also be reamed to allow for dowel movement both rotational and linear.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty