Would you like to log in again?
TeacherGeek Reamer
Supplier: TeacherGeek
1823-87
470132-526EA 7.05 USD InStock
470132-526
TeacherGeek Reamer
Educational Materials Physics Educational Materials TeacherGeek System Components
Create a pivot joint for your TeacherGeek projects.
The TeacherGeek Reamer is a tool designed by us to be compatible with our patented splined hole system. When a dowel is pressed into a hole, the spline holds for a tight fit. If a rotating or sliding fit is desired, simply turn the reamer a couple of times in the hole to remove the spline. When you’re building with TeacherGeek components, you should have it.
The TeacherGeek Reamer is one of the most useful tools in our line. It allows you to reduce or remove the spline in the holes found in connector strips, gears, wheels and perpendicular blocks. Removing the spline allows a dowel to spin freely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty