Would you like to log in again?
TeacherGeek Rubber Stretch Tires, ³/₄" wide
Supplier: TeacherGeek
1823-65
470132-624EA 26.99 USD InStock
470132-624
TeacherGeek Rubber Stretch Tires, ³/₄" wide
Educational Materials Physics Educational Materials TeacherGeek System Components
Teacher Geek Rubber Tires improve traction. Stretch these ¾" wide custom natural rubber extrusions over our Teacher Geek Wheels.
Applying glue stick on the wheel surface before installing the tires helps keep them in place. Note, these are not just average rubber bands. The tires are designed to work with our wheels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty