Would you like to log in again?
Force Probe Clamp
Supplier: VWR International
FCP-RED
470134-172EA 20.05 USD InStock
470134-172
Force Probe Clamp
Clamps General Purpose Clamps
Provide a Secure Base for Force Probe Activities.
Attach any Force Probe to a force table, inclined plane, or table edge with this easy-to-use clamp accessory. The metal clamp opens from 13 to 39 mm, and attaches securely to surfaces of all sizes. The top rod is 30 mm tall and 9.8 mm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty