Would you like to log in again?
Ward's® AP Biology Investigation 5: Cell Processes: Photosynthesis
Supplier: Avantor
Ward's®
WARD470136-828 WARD470330-120 WARD470134-770
470330-120KT 70.99 USD InStock
470330-120 470136-828 470134-770
Ward's® AP Biology Investigation 5: Cell Processes: Photosynthesis
Educational Materials Biology Educational Materials AP Biology Learning Activities
Meets AP Science Practices 1, 2, 3, 6, and 7
  • Updated illustrated instructions aid comprehension
  • Leaf pigment separation and environmental effects on photosynthetic oxygen production is quantified
  • Available for individual student use or class-size kit
In this investigation, the ability of plants to capture, store, and use energy from light for growth and reproduction is brought to life as students practice basic paper chromatography techniques to visualize pigments present in various plant leaves. Using a floating disk assay, students will investigate the environmental elements that affect the rate of photosynthesis under controlled conditions.
The Only Kits Aligned with AP Science Practices. Ward's AP Biology Investigations provide complete coverage of the Four Big Ideas and Seven Science Practices to help you meet new AP Biology curriculum standards and prepare students for real-world success in college and research-level labs. Each investigation follows a series of three parts, beginning with a structured lab investigation, followed by a related guided inquiry investigation with thoughtful assessment questions, and finally, suggestions to encourage students to formulate their own questions and explore further self-guided experimentation. This complete series of investigations emphasizes the progression to fully student-directed investigations and builds competence in all of the science practices, helping students:
• Practice concept application instead of fact memorization.
• Focus on self-guided experimentation instead of step-by-step lab procedure.
• Investigate real-world concepts with greater curriculum flexibility.
• Apply problem-solving skills to strengthen student inquiry.
AP and Advanced Placement Program are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Individual student kit contains reduced quantities of materials to perform most activities, but on an individual student basis.
Accessories information: Refill kits contain only consumable component items and do not contain additional copy masters.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty