Would you like to log in again?
Visual Scientifics Backboard
Supplier: EISCO SCIENTIFIC LLC
PTBACK
470135-558EA 88.4 USD InStock
470135-558
Visual Scientifics Backboard
Educational Materials Physics Educational Materials Mechanics Learning Activities
This backboard mounts directly to the Visual Scientifics base, enhancing video analysis.
  • Platform neutral - works with your preferred probeware/video analysis tools
  • High-Contrast Grid
  • Magnetic Backboard
  • Mounting Hardware Provided
  • Easy Installation
Students can take vertical measurements and collect data from a high-contrast grid. The backboard is magnetic and accommodates different mounting solutions for various experiments and probeware.
Mounting hardware is included. The backplace measures 2¾×16½×23½".
Ordering information: Visual Scientifics Base (470148-822) required, but sold separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty