Would you like to log in again?
Visual Scientifics Ballistic Pendulum
Supplier: EISCO SCIENTIFIC LLC
PTBALP
470135-560EA 80.5 USD InStock
470135-560
Visual Scientifics Ballistic Pendulum
Educational Materials Physics Educational Materials Mechanics Learning Activities
Students investigate the properties of momentum and conservation of energy using this precision apparatus.
  • Spring-loaded launcher, rigid-arm pendulum with barrel, and measuring board
  • Lab manual included
  • Covers NGSS and common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
The ballistic pendulum remains a staple in today's physics classrooms. This model is straightforward and easy to use. The inlcuded lab manual provides approximately three hours of laboratory activities, and meets several Next Generation Science Standards and Common Core concepts.
The pendulum is useful for demonstrating properties of momentum and energy.
Ordering information: Visual Scientifics Base (470148-822) required, but sold separately.
Delivery information: The kit includes a spring-loaded launcher, rigid-arm pendulum with barrel, measuring board, ball, and small metal rod with handle. Recommended accessories: video camera, graphing software, carbon paper, photogates, scale for obtaining masses, caliper.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty