Would you like to log in again?
Visual Scientifics Hooke's Law
Supplier: EISCO SCIENTIFIC LLC
PTHOKL PTHOKLSPRING
470332-240EA 3.05 USD InStock
470332-240 470135-562
Visual Scientifics Hooke's Law
Educational Materials Physics Educational Materials Mechanics Learning Activities
Students investigate and learn the principles of Hooke’s Law with this activity.
  • Lab manual included, with activities and extensions
  • Covers NGSS and common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
Using technology to investigate Hooke's Law, students will discover the linear relationship between applied mass and spring extension. The included lab manual provides approximately 3 hours of laboratory activities. Instructions are written to explicitly cover Next Generation Science Standards and Common Core concepts.
It's especially applicable to mechanics and physics classes.
Ordering information: Visual Scientifics Base (470148-822) required, but sold separately.
Delivery information: This kit includes 3 springs with different spring constants, aluminum support rod with hook, metal ruler, hanging mass hook with pointer, and 9 to 10 g masses. Recommended accessories: video camera and graphical analysis software.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty