Would you like to log in again?
Visual Scientifics Loop Track
Supplier: EISCO SCIENTIFIC LLC
PTLOP
470135-568EA 31.8 USD InStock
470135-568
Visual Scientifics Loop Track
Educational Materials Physics Educational Materials Mechanics Learning Activities
Students apply concepts of conservation of energy to a loop setup, then consider the forces involved in roller coasters.
  • 30-page lab manual included, with activities and extensions
  • Covers 15 NGSS and 17 common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
Students investigate the conversion between kinetic and potential energy and explore energy conservation. Using video analysis, students can investigate normal force on a roller coaster rider. The included lab manual provides approximately 3 hours of laboratory activities. Instructions are written to explicitly cover Next Generation Science Standards and common core concepts.
Ordering information: Visual Scientifics Base (470148-822) required, but sold separately. This kit includes a metal ramp with loop, and two metal balls. Recommended accessories: video camera and graphical analysis software, scale, and ruler or measuring tape.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty