Would you like to log in again?
Visual Scientifics Projectile Launcher
Supplier: EISCO SCIENTIFIC LLC
PTPROJ
470135-574EA 89.5 USD InStock
470135-574
Visual Scientifics Projectile Launcher
Educational Materials Physics Educational Materials Mechanics Learning Activities
This inquiry-based activity allows students to investigate and verify projectile equations.
  • 23 page lab manual included, with activities and extensions
  • Platform neutral - works with your preferred probeware/video analysis tools
Students explore two-dimensional projectile motion. Using video analysis with two extension activities, students verify theoretical modeling and use experimental results to meet design criteria. The included lab manual provides approximately 3 hours of laboratory activities. Instructions are written to explicitly cover Next Generation Science Standards and Common Core concepts.
Certifications: Meets 12 Next Generation Science Standards concepts and 10 common core standards.
Ordering information: Photogate system required, but sold separately (470317-428 recommended). Visual Scientifics Base (470148-822), required, but sold separately.
Delivery information: This kit includes a projectile launcher, mounting hardware, protractor with bob, and ball. Recommended accessories: video camera, graphical analysis software, carbon paper, and masking tape.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty