Would you like to log in again?
Eisco® Enlarged Upper Triple Root Molar, 2 Parts
Supplier: EISCO SCIENTIFIC LLC
Eisco®
AM0046
470136-042EA 56.9 USD InStock
470136-042
Eisco® Enlarged Upper Triple Root Molar, 2 Parts
Models Human Biology Models
An enlarged molar separated into two longitudinal sections through the crown, demonstrating the internal and external anatomy of human teeth.
  • Designed by medical professionals and educators
  • Custom molded using the highest quality nontoxic polymers and PVC
  • Includes a key card identifying nine numbered structures
  • Hand-painted by skilled artists
This hand painted molar model is enlarged 15× to show the finest details. It is separated into two longitudinal sections through the crown, two roots and pulp cavity. It comes rod-mounted on a base, with a key card identifying nine structures numbered on the model.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty