Would you like to log in again?
Eisco® Enlarged Human Teeth Models, Set of 5
Supplier: EISCO SCIENTIFIC LLC
AM0047
470136-046EA 161.4 USD InStock
470136-046
Eisco® Enlarged Human Teeth Models, Set of 5
Models Human Biology Models
Set of 5 labeled models that demonstrate the internal and extern anatomy of the 5 major human tooth types.
  • Teeth are 8 times life size
  • Hand painted
  • Each model is rod mounted on its own plastic base stand
  • Includes numbered key card
This distinct human teeth model set includes 5 unique models, illustrating each major human tooth type. The lower incisor features a removable half of the crown, the lower canine separates into two longitudinally, and the lower molar with one root is one-piece. The two root molar separates into three parts showing dental caries on various stage. The first upper molar with three roots separates into three parts. These are all hand painted with great detail.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty