Would you like to log in again?
Eisco® Color Coded Spine
Supplier: EISCO SCIENTIFIC LLC
AMCH1008AS
470136-322EA 155.39 USD InStock
470136-322
Eisco® Color Coded Spine
Models Human Biology Models
This life-size model features color-coded parts to ease research.
  • Flexible
  • Life-Size
  • Color-Coded
  • Model Includes the Sacrum, Coccyx, and More
The flexible vertebral column includes the occipital plate, cervical; thoracic and lumbar vertebrae, sacrum, coccyx, and complete pelvis. It also provides vertebral arteries, spinal nerves, and a prolapsed L3-L4 intervertebral disc.
Regions of the spinal column are color coded for identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty