Would you like to log in again?
Eisco® Life Size Female Pelvis
Supplier: Eisco Scientific
AMCHA589AS
470136-364EA 67.7 USD InStock
470136-364
Eisco® Life Size Female Pelvis
Models Human Biology Models
Useful for demonstrating the morphological distinctions between male (not included) and female pelvic structures.
  • Life sized, medical quality 3D rendering of a human female pelvic bone
  • Model shows all the predominant features of both the sacrum and coxal bones
  • Features realistic details, textures and bony landmarks
  • Incredible amount of realism for examining the human skeleton
  • Made of durable and sturdy pvc plastic
  • Washable model is easy to clean and will hold up to years of handling
Life size model of female pelvis. Includes the left and right innominates with pubic symphysis. Ideal for studying the structure and function of this part of the skeletal system, and is an excellent tool for comparing male and female skeletal anatomy. Incredible amount of realism and detail. Model shows all the predominant features of the sacrum (median sacral crest, lateral sacral crests, posterior sacral foramina, ala and sacral hiatus) and the coxal bones (iliac crest, pelvic brim, ischial spine, acetabulum, pubic crest, pubic symphysis, ischium, pubic bone, and illium). Realistic textures and bony landmarks make this replica an excellent tool for educational study, as well as for patient demonstrations. Made of durable, sturdy, and washable pvc plastic, this replica will withstand long term use and handling. Model measures approximately 10.5" tall, 9" wide, and 5" long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty