Would you like to log in again?
Clinical Diagnosis of Diabetes Kit
Supplier: Edvotek
280
470136-466KT 203.1 USD InStock
470136-466
Clinical Diagnosis of Diabetes Kit
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
Over 380 million people worldwide are afflicted by diabetes mellitus, a chronic disease that leads to high blood sugar. Due to genetic predisposition and high-calorie, low-activity lifestyles, that number continues to grow. Without early detection and treatment of diabetes, severe medical complications can occur. In this simulation, students will diagnose diabetes in three patients using the urine glucose test and Enzyme-linked Immunosorbent Assay (ELISA).
Note: If shipping to warehouse, perishable coupon included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty