Would you like to log in again?
Detection of GMOs Kit
Supplier: Edvotek
121
470136-468KT 140.75 USD InStock
470136-468
Detection of GMOs Kit
Educational Materials Biology Educational Materials Genetics Learning Activities
For centuries, humans have used selective breeding and conventional hybridization to produce desirable qualities and to increase crop yields. Today, scientists use genetic engineering to directly manipulate the DNA, quickly producing these desirable traits. In this experiment, students will use agarose gel electrophoresis to explore the molecular methods used by scientists to identify genetically modified organisms. No thermal cycler is required. Students are also encouraged to explore the controversy surrounding the use of genetically modified organisms.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty