Would you like to log in again?
Ward's® Chemistry Hydrogen Fuel Cell Demonstration
Supplier: Ward's
IS9507
470136-728KT 60.7 USD InStock
470136-728
Ward's® Chemistry Hydrogen Fuel Cell Demonstration
Educational Materials Chemistry Educational Materials Green Chemistry Learning Activities
Show How Energy is Produced.
  • Green chemistry
  • Understand catalysts
  • All chemicals required are included
Your students will be amazed at how easily energy can be produced by combining hydrogen and oxygen to form water. In this demonstration, platinum serves as the catalyst, and electrodes will be prepared by coating metal mesh with the platinum. The hydrogen and oxygen come from electrolysis. After the cell is set up, a brief current is applied with a nine-volt battery, causing the formation of hydrogen gas bubbles on one electrode, and oxygen gas bubbles on the other. Using a voltmeter, electricity produced by the recombining of hydrogen and oxygen, facilitated by the platinum metal catalyst, can be observed. 
Ordering information: Includes Brass Metal Mesh Squares, Red Connecting Wires with Alligator Clips, Black Connecting Wires with Alligator Clips, D-Cell Battery Holder with Clips, Chloroplastinic Acid, Glass Rod, and Instructions. Required but Not Included: Voltmeter, 9-Volt Battery, Basic Glassware/Labware.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty