Would you like to log in again?
LaMotte® BioPaddles
Supplier: LaMotte
5550 5846 5552 5551 5553
470136-862EA 57.6 USD InStock
470136-862 470136-846 470136-848 470136-864 470136-844
LaMotte® BioPaddles
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Students identify and quantify microbes in air, soil, water, or any surface.
  • Easy to Use
  • No Refrigeration Needed
  • Ten Dual-Agar Paddles
  • App Included
  • Technical Information Sheet Provided
These ready-to-use dual-agar paddles have a longer shelf life than traditional Petri dishes. Each BioPaddle contains microbe-specific media enclosed in a sterile vial. The paddles do not require any other testing equipment. Only a magnifier and warm place (35°C or an incubator) are needed for air, soil, liquid, or surface sampling.
All BioPaddle products include a free app that lets students compare the unknown microbe growth on the paddle to known microbe colonies from six microhabitats shown in a library of photos. The app also has the option of capturing the BioPaddle image for side-to-side comparison with library photos.
Ordering information: This kit includes ten paddles per box and a technical information sheet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty