Would you like to log in again?
Earth's Interior Globe
Supplier: VWR International
BD0078
470136-882EA 248.95 USD InStock
470136-882
Earth's Interior Globe
Globes
Get an Inside Look at Our World
Every student will be in awe of our planet with this beautifully hand-painted resin model in your classroom. It depicts the internal structure of the earth, featuring seven detachable parts that illustrate the crust, mantle, lithosphere, mesosphere, asthenosphere, and inner and outer core. Size: 9" diameter globe, mounted on a sturdy metal tripod base.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty