Would you like to log in again?
NeuLog Light Logger Sensor
Supplier: EISCO SCIENTIFIC LLC
NUL204
470137-376EA 79.5 USD InStock
470137-376
NeuLog Light Logger Sensor
Data Loggers
This adaptable light sensor has a wide range of uses.
  • Three ranges for illumination measurement
  • Fast and slow modes available
  • Ideal in low-light and high-light environments
It can be used to study photosynthesis in biology, light-emitting chemical reactions in chemistry, the effect of changing voltage on a light-bulb's output in physics, and more. This sensor measures illumination with three ranges.
With both fast and slow modes, it can measure fast light changes, such as those produced by light bulbs connected to an AC supply, and the almost-steady levels outside on a sunny day.
It supports experiments that last from 50 ms to 31 days.
Ordering information: One or more of the following items are needed but not included–a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand-alone probe or if you have the wireless module), wireless module (not needed if connecting to a computer through the USB module).
Caution: For educational use only.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty