Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NeuLog Sound Logger Sensor
Supplier: EISCO SCIENTIFIC LLC
NUL212
470137-392EA 119.5 USD InStock
470137-392
NeuLog Sound Logger Sensor
Data Loggers
Two measuring modes, many uses
  • Slow and fast modes
  • Compare wavelengths
  • Determine frequencies
  • Maximum sample rate of 100 S/s, 10000 S/s (signal)
With both a slow measuring mode for decibels and a fast mode for comparing different wavelengths of sound this sensor supports experiments lasting between 25 milliseconds and 31 days. Using the two sound sensors the velocity of sound propagation is easily determined. Determine frequencies of objects such as tuning forks and windchimes or use it to calibrate simple electronic signal generators.
For each probe you will need one or more of the following items: Battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WiFi module), WiFi module (not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty