Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NeuLog Force Plate Logger Sensor
Supplier: EISCO SCIENTIFIC LLC
NUL225
470137-416EA 362.85 USD InStock
470137-416
NeuLog Force Plate Logger Sensor
Data Loggers
This sensor helps you tackle heavy measurements.
  • For Experiments Lasting between One Second and 31 Days
  • Ideal for More Difficult Measurements
  • Battery Module USB Module Recommended
It supports experiments lasting between One second and 31 days.
The sensor is ideal for more difficult measurements.
Ordering information: The following items are needed but not included: a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if using as a stand-alone probe or you have the Wi-Fi module), and Wi-Fi module (not needed if connecting to a computer through the USB module). A battery module and a USB module are recommended.
Caution: For educational use only.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty