Would you like to log in again?
NeuLog Soil Moisture Logger Sensor
Supplier: EISCO SCIENTIFIC LLC
NUL229
470137-424EA 148.05 USD InStock
470137-424
NeuLog Soil Moisture Logger Sensor
Data Loggers
A unique approach to measuring moisture via vacuum pressure in a tensiometer.
  • Max. sample rate 100 S/sec.
  • Experimental duration 1 sec. to 31 days
The tensiometer on this logger sensor measures soil moisture via pressure level. When the soil is dry the water exits ceramic holes in the end due to diffusion, when the soil is wet the vacuum in the tensiometer pulls water inside and the vacuum decreases.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty