Would you like to log in again?
Optics Bench Kit
Supplier: EISCO SCIENTIFIC LLC
PH0649
470137-548EA 161.7 USD InStock
470137-548
Optics Bench Kit
Educational Materials Physics Educational Materials Optics Learning Activities
Study optics with this complete kit while learning about light, refraction, reflection, convex and concave lenses, and focal length.
This enhanced set of accessories allows users to perform a wide variety of experiments in optics.
The kit includes: a bench, riders with tightening screws, a lamp with halogen bulb, lens holders, a variety of lenses, set of diaphragms, a mirror, and various screens.
Ordering information: Power supply required; not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty