Would you like to log in again?
Solar Panels
Supplier: TEACHERGEEK INC
1824-10 1824-09
470137-614PK 403.49 USD InStock
470137-614 470137-612
Solar Panels
Educational Materials Earth Science Educational Materials Astronomy Learning Activities
This solar panel features a PET backed panel with alligator clip leads.
  • Lightweight
It can mount on projects such as the solar racer, boat or any other invention. It should output plenty of power for your projects: 3 Watts, 6 Volts, .5 Amps and works with large 3-6V motor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty