Would you like to log in again?
Student Coin and Feather Tube
Supplier: VWR International
TGF-SPL12
470137-620EA 115.2 USD InStock
470137-620
Student Coin and Feather Tube
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
The Student Coin and Feather Tube shows the two items in a vacuum, falling at the same rate.
  • Tube Length: 90 cm
  • Rubber End Caps Connect to Tube
  • Feather and Disk Included
  • Economical
  • Hose Cock Connects to Any Standard Vacuum Pump
A transparent plastic tube, 90 cm long and 4.5 cm in diameter, features rubber end caps.
One end has a hose and hose cock that hooks up to any standard vacuum pump to remove air from the tube.
Ordering information: This kit includes a feather and disk. A vacuum pump is required but not provided.
Caution: Caution:This product has small parts. It is not for children under three years. CHOKING HAZARD.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty