Would you like to log in again?
Franck-Hertz Experiment Power Supply
Supplier: AMERICAN 3B SCIENTIFIC SE
1012818
470137-702EA 2174.39 USD InStock
470137-702
Franck-Hertz Experiment Power Supply
Power Supplies
Adjustable Voltage Power Supply
This is the power supply unit for operating the mercury-filled Franck-Hertz tube and the neon-filled Franck-Hertz tube. It provides all the voltages needed to power the tubes and includes a sensitive built-in DC amplifier for measuring collector current. The accelerating voltage can be set-up manually on the apparatus or set to a saw-tooth wave form. Additional measuring inputs are also available for the anode current and accelerating voltage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty