Would you like to log in again?
Spherometer
Supplier: United Scientific Supplies
SPRM01
470137-716EA 39.35 USD InStock
470137-716
Spherometer
Educational Materials Physics Educational Materials Optics Learning Activities
Precision Measurement of Very Small Lengths
  • Easy to read dials
  • Accurate to 0.01 mm
This useful instrument determines the radius of curvature of spherical surfaces such as mirrors and lenses. Its micrometer screw is graduated to measure displacement as small as 0.01mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty