Would you like to log in again?
Multipurpose Clamp Holder
Supplier: EISCO SCIENTIFIC LLC
CH0663D
470137-830EA 12.7 USD InStock
470137-830
Multipurpose Clamp Holder
Clamps General Purpose Clamps
Clamp to Almost Any Surface
This multipurpose clamp holder accommodates both square and round rods and can be clamped to table edges, other rods, boards and more. It is made of aluminum alloy with plastic head thumbscrews. This clamp has the ability to accommodate objects up to 16mm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty