Would you like to log in again?
Suspension Clamp
Supplier: EISCO SCIENTIFIC LLC
CH0667
470137-844EA 10.75 USD InStock
470137-844
Suspension Clamp
Clamps General Purpose Clamps
Takes the Heat
Use this suspension clamp to safely hold a thermometer or specimens over a hotplate. The clamp keeps objects a distance of 110mm from the rod and the hook can be adjusted to any angle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty