Would you like to log in again?
Stand and Rod Deluxe Pack
Supplier: EISCO SCIENTIFIC LLC
STDRD1
470137-894EA 37.75 USD InStock
470137-894
Stand and Rod Deluxe Pack
Support Stands
Solid and Sturdy
The base in this pack is base cut steel and is a heavier choice than the standard version. The powder-coated base has rubber backing to prevent slipping. Rod diameter is 12mm and they thread directly into the base.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty