Would you like to log in again?
Eisco® Root Tip Model
Supplier: EISCO SCIENTIFIC LLC
BM0029
470138-616EA 192.99 USD InStock
470138-616
Eisco® Root Tip Model
Models Botany Models
This model of root tip shows the structure of a typical monocot root in a cross section and longitudinal section.
  • Mounted on stand with numbered key card
Root structure is easily examined including the root cap, root hairs, zone of distention, meristematic zone and zone of primary structure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty