Would you like to log in again?
Low Friction Pulley With Rod
Supplier: EISCO SCIENTIFIC LLC
PH0300-SPL
470138-852EA 27.2 USD InStock
470138-852
Low Friction Pulley With Rod
Educational Materials Physics Educational Materials Mechanics Learning Activities
Total length of rod and pulley is 10". The pulley has a diameter of 2", and is equipped with a metal low-friction bearing installed. The rod is made of durable aluminum, and the pulley is made of durable plastic.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty