Would you like to log in again?
Lattice Rod, Aluminum
Supplier: EISCO SCIENTIFIC LLC
CH0658PLM3 CH0658PLM6
470140-276EA 16.15 USD InStock
470140-276 470140-274
Lattice Rod, Aluminum
Support Rods and Frames
Resists corrosion
  • Made of T6 Temper 6063 aluminum alloy
These anodized, smooth, unthreaded aluminum rods are used for constructing lattice rod assemblies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty