Would you like to log in again?
Heavy Support Stands with Rods, Stamped Steel
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SSB4X6-H
470144-202EA 27.1 USD InStock
470144-202
Heavy Support Stands with Rods, Stamped Steel
Support Stands
Sturdy and heavy support stands.
  • Extra heavy base
  • Thicker screw hole mounts
  • Includes a support rod
Heavy stamped steel bases with black enamel finish feature an added steel plate on the back, which makes them about 50% heavier than regular stamped steel bases. The steel plate also facilitates a sturdier threading of the rod into the base.
Delivery information: Includes a removable, zinc-plated steel rod.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty