Would you like to log in again?
Whirlybird™ Wind Turbine Kit
Supplier: PICOSOLUTIONS GROUP INC SE
WBWTK
470144-252CS 3074.5 USD InStock
470144-252
Whirlybird™ Wind Turbine Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
For Indoor and Outdoor Use.
Use the simple components of a common roof ventilator to create your own 3-phase wind turbine. All parts are included to build a commercial-grade wind power device using common household tools, and without the need for soldering or drilling. Use your computer and the included 3-phase Bar Graph Board to plot and analyze wind patterns and corresponding electrical output. Ideal for both indoor and outdoor use. Durable bearings allow the turbine to stand up to even the most severe wind and weather conditions, enabling students to study day-to-day wind patterns all year long.
Please Note: Roof Ventilator not included. We suggest the Lomanco Whirlybird™ model B1B-12 roof ventilator purchased separately at home improvement outlets.
Included Hardware:
12 Wire Coils
16 Bar magnets
1 6-position terminal block
1 8” diameter round metal rotor disk
1 8” diameter round Plexiglas stator disk
1 5/16”, 7/8” long threaded rod extender
1 5/16”, 1” long bolt
10 5/16” flat washers
3 1" flat washers
  • No soldering or drilling required
  • Durable components function year round
  • Build a long term data set
Use the simple components of a common roof ventilator to create your own 3-phase wind turbine. All parts are included to build a commercial-grade wind power device using common household tools, and without the need for soldering or drilling. Use your computer and the included 3-phase Bar Graph Board to plot and analyze wind patterns and corresponding electrical output. Ideal for both indoor and outdoor use. Durable bearings allow the turbine to stand up to even the most severe wind and weather conditions, enabling students to study day-to-day wind patterns all year long.
Please Note: Roof Ventilator not included. We suggest the Lomanco Whirlybird™ model B1B-12 roof ventilator purchased separately at home improvement outlets.
Included Hardware:
Wire Coils
Bar magnets
6-position terminal block
8” diameter round metal rotor disk
8” diameter round Plexiglas stator disk
5/16”, 7/8” long threaded rod extender
5/16”, 1” long bolt
5/16” flat washers
1"" flat washers
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty