Would you like to log in again?
Art Forgery Kit
Supplier: MASTERDATA QL HOLD 2
10-524
470144-258CS 1249.15 USD InStock
470144-258
Art Forgery Kit
Educational Materials Chemistry Educational Materials Consumer Chemistry Learning Activities
Students compare real paintings from forgeries.
  • Grades 6-12
  • Use scientific observation and reasoning
Kemtec Art in Chemistry; Chemistry in Art has 4 units which can be used individually. Instead of purchasing the full student kit for all units, now you can infuse these activities into your curriculum using these unit kits. Each kit includes materials and expendable chemicals for 6 lab groups, a group having up to 4 students.
New kits include:
Detection of an Art Forgery-Use chemical and ultraviolet lamp tests to determine which paintings are forgeries. Includes 5 pairs of paintings.
Making Paints: The Electromagentic Spectrum-Explore the visible spectrum and the uses of pigments.
Electrochemistry and Jewelry Making-Using jewelry, explore metallic properties and how to electroplate materials.
Photography/Redox, Solubility, Equilibrium-Build a simple camera and explore the chemical processes in developing and printing a photo.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty