Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Kemtec® Introduction to Biochemistry 2

Supplier: MASTERDATA QL HOLD 2

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

2-217
470144-272CS 2251.5 USD InStock
470144-272
Kemtec® Introduction to Biochemistry 2
Educational Materials Biology Educational Materials Biology Textbooks
A more in-depth biochemical analysis of foods.

  • Ideal for biology, nutrition, general science, and biochemistry courses
  • All required chemicals included
  • Target grades 9-12

Expose upper level students to an expansive survey of fascinating biochemistry experiments. The kit introduces students to scientific techniques while examining ten different topics at a more in-depth level than Biochemistry I (363000). Includes: determining the pH of foods, saliva, and soil; starch determination; sugar determination; fat determination; protein determination; amino acid determination; pepsin to digestion of protein; ninhydrin amino acid detection; effects of varying conditions on salivary amylase activity; modeling how aigestion works; determination of ascorbic acid concentration; and paper chromatography of various dyes and plant pigments using different solvents. Designed for 24 students working in six groups of four.

Includes:
Instructor Manual
SDS
Methods of Assessment Manual
Chromatography Paper
pH Paper 1-14
Filter Paper
Dialysis Tubing
Snap Caps
8 Dram Shell Vials
Pipettes
Micro Pipettes
Empty Bottles
Benedict's Solution
Biuret Solution
Hydrochloric Acid Solution
Sodium Hydroxide Solution
Starch
Glucose
Protein
Lipase
Pepsin
Teacher's Guide and Student Copymaster

Required but Not Included: A hot plate or water bath, 50 mL beakers, forceps, storage jars with caps, funnels, and food samples for testing.

Delivery information: See datasheet for contents information.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty