Would you like to log in again?
Glass Prism and Lens Set with Storage Case
Supplier: United Scientific Supplies
OPSETG2
470144-438EA 139.35 USD InStock
470144-438
Glass Prism and Lens Set with Storage Case
Educational Materials Physics Educational Materials Optics Learning Activities
High Quality, Scratch Resistant Set
  • Made of smooth, polished glass for durability and brilliant transmission of light
  • The set is housed in a protective case
Use this seven-piece set of superior prisms and lenses to teach a whole range of optical principles. Includes the following: 45° and 60° trapezoidal prism, equilateral prism, two right-angle prisms (60mm high, 15mm hypotenuse, 10mm sides; and 51mm high, 32mm hypotenuse, 24mm sides), a double-concave and a double-convex lens that may be used together, and a semicircular lens.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty