Would you like to log in again?
LED Light Engines
Supplier: DIGITRON ELECTRONICS SE
LHA06B12BSB
470144-580PK 17.05 USD InStock
470144-580
LED Light Engines
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Examine the Role of Light Waves in Alternative Energy
  • Versatile
  • Thought-Provoking
In this experiment, students will compare the energy efficiency of LED, incandescent, and compact fluorescent lights when tested utilizing standard, wind, and solar power sources. Six Ultra Brite LEDs produce high output with efficiency of 150ma at 12V. The plug and play design allows students to connect the Light Engines directly to a power source. Unlike 1–watt surface mount LEDs, the 1–watt LED light engines do not require a heat sink. Designed for use with regulated power supply or external LED driver module.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty