Would you like to log in again?
Precision Measuring Set
Supplier: United Scientific Supplies
Avg. Ratings: 5.0 (1 Review)
REVIEWS (1)
Total Ratings: 1
Avg. Ratings: out of 5
Sort all by:
PMSET04
470144-808PK 185.25 USD InStock
470144-808
Precision Measuring Set
Educational Materials Math Educational Materials Computation/Estimation Learning Activities
Four High-Quality Instruments Housed in a Sturdy Plastic Storage Case.
  • Complete kit is supplied in a fitted plastic case for easy stacking
  • Transparent top allows for easy inspection
  • Instructions included
Teach precision linear measurement, significant figures, and approximation with this unique kit. Give your students concrete experience using industrial tools and various unknowns. The kit includes a detailed instruction manual, a micrometer, vernier caliper, a knife-edge square, and a ruler. Unknowns to be measured include a 3/4" steel sphere, a round aluminum rod, a plugged aluminum tube, and a rectangular aluminum block.
Kit Contains:
1 Micrometer Caliper, 0-25mm x 0.01mm
1 Knife-Edge Square, 90°, 100mm x 70m
1 Vernier Caliper, 150mm x 0.02mm
1 Ruler, Steel, 15cm x 0.5mm
Order Now
ORDER
WRITE A REVIEW
REVIEWS (1)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty