Would you like to log in again?
Screw Compressor Clamp with Closed Jaw
Supplier: United Scientific Supplies
HSCCL1
470144-828CS 36.85 USD InStock
470144-828
Screw Compressor Clamp with Closed Jaw
Clamps General Purpose Clamps
Used to control flow in flexible tubing.
  • Fits tubes up to 19 mm
  • Easy to use thumbscrew
This screw compressor clamps and holds flexible tubes securely closed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty