Would you like to log in again?
DNA DuraGel™
Supplier: Edvotek
S-43-20 S-43
470145-082PK 48.95 USD InStock
470145-082 470145-002
DNA DuraGel™
Electrophoresis Gels Agarose and Acrylamide
Ideal for student skill practice.
  • Durable, reusable practice gels
  • Accurately replicate actual agarose gel
  • Detailed instructions included
DNA DuraGel™ gels are permanent polymer gels that allow students to practice the critically important skill of pipetting/gel loading. The clear, reusable gels are designed for the practice of loading 5 – 35 µL of samples. The gel grids are imprinted with a ruler for sizing DNA fragments. Also included are simulated FlashBlue™ and Ethidium Bromide gel images, ideal for representing how actual gels are stained with Methylene Blue and Ethidium Bromide. Pipetting DNA and biological reagents is a critical skill required for biotechnology and life science experiments. This DNA DuraGel™ is designed for proper student training, to teach techniques for pipetting samples for agarose gel electrophoresis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty