Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Disposable Thermometers
Supplier: MEDICAL INDICATORS,INC
1112-20
470145-068CS 802.9 USD InStock
470145-068
Disposable Thermometers
Thermometers Analog Thermometers
These affordable, single-use thermometers are accurate yet eliminate the need for sterilization between uses.
  • For Oral or Axillary Readings
  • Individually Wrapped
  • Range: 96 to 104.8°F
The last dot on the scale to turn from green to black indicates the correct temperature, and the results work for either oral or axillary readings.
The mercury-free, individually wrapped thermometers have a range of 96.0 to 104.8°F in 0.2° increments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty